Thermo Scientific Chemicals

Methyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: H61758.22
100 g, Each
Thermo Scientific Chemicals

Methyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: H61758.22
100 g, Each
Quantity
Catalog number: H61758.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
333-27-7
Proton NMR
97.5% min

Description

Methyl trifluoromethanesulfonate is used as a powerful methylating reagent in chemistry. Further, it is used in the conversion of amines to methyl ammonium triflates. In addition, it is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trifluoromethanesulfonate is used as a powerful methylating reagent in chemistry. Further, it is used in the conversion of amines to methyl ammonium triflates. In addition, it is also used in Suzuki reaction.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases, strong acids and strong reducing agents. Hydrolyzes with water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text