2-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals
2-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number H61785.14
also known as H61785-14
Price (USD)/ Each
282.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
282.00
Add to cart
2-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals
Catalog numberH61785.14
Price (USD)/ Each
282.00
-
Add to cart
Chemical Identifiers
CAS59907-13-0
IUPAC Name2-bromo-1-fluoro-3-methylbenzene
Molecular FormulaC7H6BrF
InChI KeyFYCXRRYRNRDSRM-UHFFFAOYSA-N
SMILESCC1=CC=CC(F)=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
2-Bromo-3-fluorotoluene is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluorotoluene is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only