Thermo Scientific Chemicals

2-(4-Fluorophenyl)ethylamine, 97%, Thermo Scientific Chemicals

Catalog number: H61791.22
100 g, Each
Thermo Scientific Chemicals

2-(4-Fluorophenyl)ethylamine, 97%, Thermo Scientific Chemicals

Catalog number: H61791.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H61791.22
also known as H61791-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1583-88-6
IUPAC Name
2-(4-fluorophenyl)ethan-1-aminium
Molecular Formula
C8H11FN
InChI Key
CKLFJWXRWIQYOC-UHFFFAOYSA-O
SMILES
[NH3+]CCC1=CC=C(F)C=C1
Assay (GC)
>96.0%

Description

2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text