Thermo Scientific Chemicals

5-Fluoro-2-nitroaniline, 97%, Thermo Scientific Chemicals

Catalog number: H61806.06
5 g, Each
Thermo Scientific Chemicals

5-Fluoro-2-nitroaniline, 97%, Thermo Scientific Chemicals

Catalog number: H61806.06
5 g, Each
Quantity
Catalog number: H61806.06
also known as H61806-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2369-11-1
IUPAC Name
5-fluoro-2-nitroaniline
Molecular Formula
C6H5FN2O2
InChI Key
PEDMFCHWOVJDNW-UHFFFAOYSA-N
SMILES
NC1=CC(F)=CC=C1[N+]([O-])=O
Assay (HPLC)
>96.0%

Description

5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text