5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
Jong-Beom Baek; and Frank W. Harris. Poly(arylether amides) and poly(aryletherketone amides) via aromatic nucleophilic substitution reactions of self-polymerizable AB and AB2 monomers. Journal of Polymer Science Part A: Polymer Chemistry. 2003, 41(15), 2374-2389.
J. C. Arcos; J. A. Miller. Synthesis of 2-Amino-5-dimethylaminodiphenylamine and Other Derivatives of 3,4-Dinitrodimethylaniline. J. Am. Chem. Soc. 1955, 77(11), 3128-3130.