Chemical Identifiers
CAS14588-08-0
IUPAC Name(acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane)
Molecular FormulaC40H36O4P2Pd
InChI KeyYOUIUHDWQIUKAO-UHFFFAOYSA-L
SMILESCC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more