Thermo Scientific Chemicals

Methyl 5-bromo-2-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H61861.06
5 g, Each
Thermo Scientific Chemicals

Methyl 5-bromo-2-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H61861.06
5 g, Each
Quantity
Catalog number: H61861.06
also known as H61861-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79669-50-4
IUPAC Name
methyl 5-bromo-2-methylbenzoate
Molecular Formula
C9H9BrO2
InChI Key
FDCYLMYCHALQJR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Br)=CC=C1C
Assay (HPLC)
>97.5%

Description

Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-bromo-2-methylbenzoate is used as a catalytic agent, petrochemical additive

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text