Specifications
Chemical Name or Material3-Bromo-2-cyanobenzeneboronic acid
CAS1032231-32-5
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more