Thermo Scientific Chemicals

6-Chloroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H61911.06
5 g, Each
Thermo Scientific Chemicals

6-Chloroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H61911.06
5 g, Each
Quantity
Catalog number: H61911.06
also known as H61911-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
703-82-2
IUPAC Name
6-chloro-1H-indole-3-carbaldehyde
Molecular Formula
C9H6ClNO
InChI Key
CTNIXLBHXMSZKL-UHFFFAOYSA-N
SMILES
ClC1=CC=C2C(NC=C2C=O)=C1
Assay (HPLC)
>97.5%

Description

6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text