3,5-Dimethylisoxazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
3,5-Dimethylisoxazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethylisoxazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H61912.14
also known as H61912-14
Price (USD)/ Each
532.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
532.00
Add to cart
3,5-Dimethylisoxazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH61912.14
Price (USD)/ Each
532.00
-
Add to cart
Chemical Identifiers
CAS54593-26-9
IUPAC Name3,5-dimethyl-1,2-oxazole-4-carbaldehyde
Molecular FormulaC6H7NO2
InChI KeyTVAYXKLCEILMEA-UHFFFAOYSA-N
SMILESCC1=C(C=O)C(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only