4-Thiouracil, 97%, Thermo Scientific Chemicals
4-Thiouracil, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Thiouracil, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H61919.03
also known as H61919-03
Price (USD)/ Each
94.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
94.40
Add to cart
4-Thiouracil, 97%, Thermo Scientific Chemicals
Catalog numberH61919.03
Price (USD)/ Each
94.40
-
Add to cart
Chemical Identifiers
CAS591-28-6
IUPAC Name4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
Molecular FormulaC4H4N2OS
InChI KeyOVONXEQGWXGFJD-UHFFFAOYSA-N
SMILESO=C1NC=CC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only