Thermo Scientific Chemicals

2-Amino-3-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H61925.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-3-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H61925.06
5 g, Each
Quantity
Catalog number: H61925.06
also known as H61925-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20776-51-6
IUPAC Name
2-amino-3-bromobenzoate
Molecular Formula
C7H5BrNO2
InChI Key
SRIZNTFPBWRGPB-UHFFFAOYSA-M
SMILES
NC1=C(Br)C=CC=C1C([O-])=O
Assay (HPLC)
>96.0%

Description

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text