2,5-Dibromo-3-nitropyridine, 98%, Thermo Scientific Chemicals
2,5-Dibromo-3-nitropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dibromo-3-nitropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H61961.22
also known as H61961-22
Price (USD)/ Each
640.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
640.00
Add to cart
2,5-Dibromo-3-nitropyridine, 98%, Thermo Scientific Chemicals
Catalog numberH61961.22
Price (USD)/ Each
640.00
-
Add to cart
Chemical Identifiers
CAS15862-37-0
IUPAC Name2,5-dibromo-3-nitropyridine
Molecular FormulaC5H2Br2N2O2
InChI KeyOQKWPJCAKRVADO-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Br)=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only