2-Fluoro-4-iodobenzonitrile, 98%, Thermo Scientific Chemicals
2-Fluoro-4-iodobenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-4-iodobenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61973.03
also known as H61973-03
Price (USD)/ Each
123.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
123.00
Add to cart
2-Fluoro-4-iodobenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberH61973.03
Price (USD)/ Each
123.00
-
Add to cart
Chemical Identifiers
CAS137553-42-5
IUPAC Name2-fluoro-4-iodobenzonitrile
Molecular FormulaC7H3FIN
InChI KeyWHQVXHBSTRFRCE-UHFFFAOYSA-N
SMILESFC1=C(C=CC(I)=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only