4-Bromo-3-nitrobenzoic acid, 95%, Thermo Scientific Chemicals
4-Bromo-3-nitrobenzoic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61975.06
also known as H61975-06
Price (USD)/ Each
60.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
60.90
Add to cart
4-Bromo-3-nitrobenzoic acid, 95%, Thermo Scientific Chemicals
Catalog numberH61975.06
Price (USD)/ Each
60.90
-
Add to cart
Chemical Identifiers
CAS6319-40-0
IUPAC Name4-bromo-3-nitrobenzoic acid
Molecular FormulaC7H4BrNO4
InChI KeyRVCTZJVBWNFYRU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only