Thermo Scientific Chemicals

5'-Bromo-2'-hydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: H61979.18
50 g, Each
Thermo Scientific Chemicals

5'-Bromo-2'-hydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: H61979.18
50 g, Each
Quantity
Catalog number: H61979.18
also known as H61979-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1450-75-5
IUPAC Name
1-(5-bromo-2-hydroxyphenyl)ethan-1-one
Molecular Formula
C8H7BrO2
InChI Key
HQCCNFFIOWYINW-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC(Br)=CC=C1O
Assay (HPLC)
>97.5%

Description

5?-Bromo-2?-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2?-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N?-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

Solubility
Soluble in Chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text