5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
Leonard R. MacGillivray.; Jennifer L. Reid.; John A. Ripmeester. Supramolecular Control of Reactivity in the Solid State Using Linear Molecular Templates. J. Am. Chem. Soc. 2000, 122, (32), 7817-7818.
Derek H.R. Barton.; Dervilla M.X. Donnelly.; Jean-Pierre Finet.; Patrick J. Guiry. Total synthesis of isorobustin. Tetrahedron Letters. 1990, 31, (51), 7449-7452.