Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid 4-benzyl ester, 97%, Thermo Scientific Chemicals

Catalog number: H62011.14
25 g, Each
Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid 4-benzyl ester, 97%, Thermo Scientific Chemicals

Catalog number: H62011.14
25 g, Each
Quantity
Catalog number: H62011.14
also known as H62011-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86060-84-6
IUPAC Name
4-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Molecular Formula
C26H23NO6
InChI Key
OQGAELAJEGGNKG-UHFFFAOYNA-N
SMILES
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>96.0%

Description

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text