N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%, Thermo Scientific Chemicals
N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62018.06
also known as H62018-06
Price (USD)/ Each
195.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
195.00
Add to cart
N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%, Thermo Scientific Chemicals
Catalog numberH62018.06
Price (USD)/ Each
195.00
-
Add to cart
Specifications
Chemical Name or MaterialN,N'-Di-Boc-L-histidine dicyclohexylammonium salt
Melting Point160°C to 165°C (decomposition)
CAS31687-58-8
ColorWhite
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only