Nalpha,Nepsilon-Di-Boc-L-lysine, 98%, Thermo Scientific Chemicals
Nalpha,Nepsilon-Di-Boc-L-lysine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha,Nepsilon-Di-Boc-L-lysine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H62019.14
also known as H62019-14
Price (USD)/ Each
60.65
Online exclusive
67.00 
Save 6.35 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.00 
Save 6.35 (9%)
Add to cart
Nalpha,Nepsilon-Di-Boc-L-lysine, 98%, Thermo Scientific Chemicals
Catalog numberH62019.14
Price (USD)/ Each
60.65
Online exclusive
67.00 
Save 6.35 (9%)
-
Add to cart
Chemical Identifiers
CAS2483-46-7
IUPAC Name(2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid
Molecular FormulaC16H30N2O6
InChI KeyFBVSXKMMQOZUNU-LDGXTIHJNA-N
SMILESCC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
FormSolid
Assay from Suppliers CofA≥96.0% (HPLC)
Optical Rotation-2.75° to -3.75° (c=5 in methanol)
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

Solubility
Soluble in chloroform, ether, ethyl acetate and methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only