Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62024.MD
also known as H62024-MD
Price (USD)/ Each
166.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
166.00
Add to cart
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH62024.MD
Price (USD)/ Each
166.00
-
Add to cart
Specifications
Chemical Name or MaterialMethyl trans-3-aminocyclohexanecarboxylate hydrochloride
CAS712313-64-9
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC8H16ClNO2
View more
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only