trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H62034.06
also known as H62034-06
Price (USD)/ Each
872.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
872.00
Add to cart
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
Catalog numberH62034.06
Price (USD)/ Each
872.00
-
Add to cart
Specifications
Chemical Name or Materialtrans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine
CAS177582-74-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only