2-Bromo-6-fluorobenzylamine hydrochloride, 96%, Thermo Scientific Chemicals
2-Bromo-6-fluorobenzylamine hydrochloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-6-fluorobenzylamine hydrochloride, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62086.06
also known as H62086-06
Price (USD)/ Each
800.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
800.00
Add to cart
2-Bromo-6-fluorobenzylamine hydrochloride, 96%, Thermo Scientific Chemicals
Catalog numberH62086.06
Price (USD)/ Each
800.00
-
Add to cart
Specifications
Chemical Name or Material2-Bromo-6-fluorobenzylamine hydrochloride
CAS1214330-90-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only