trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Yongfeng Li; Shuli Mao; Michael W. Hager; Kimberlynne D. Becnel; Raymond F. Schinazi; Dennis C. Liotta. Synthesis and evaluation of 2'-substituted cyclobutyl nucleosides and nucleotides as potential anti-HIV agents. Bioorganic & Medicinal Chemistry Letters. 2008, 17(12), 3398-3401.
- Vijay Kaiwar; Colin B. Reese; Emily J. Gray; and Stephen Neidle. Synthesis of 9-[cis-3-(hydroxymethyl)cyclobutyl]-adenine and -guanine. J. Chem. Soc., Perkin Trans. 1. 1995, (18), 2281-2287.