Chemical Name or Material4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester
CAS1220219-61-3
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.