Thermo Scientific Chemicals

Nalpha-Boc-Nepsilon-Fmoc-D-lysine, 95%, Thermo Scientific Chemicals

Catalog number: H62116.06
5 g, Each
Thermo Scientific Chemicals

Nalpha-Boc-Nepsilon-Fmoc-D-lysine, 95%, Thermo Scientific Chemicals

Catalog number: H62116.06
5 g, Each
Quantity
Catalog number: H62116.06
also known as H62116-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115186-31-7
IUPAC Name
(2R)-2-{[(tert-butoxy)carbonyl]amino}-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular Formula
C26H32N2O6
InChI Key
JYEVQYFWINBXJU-ANBDAQEENA-N
SMILES
CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>94.0%

Description

Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text