Thermo Scientific Chemicals

L-Threonine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H62118.03
1 g, Each
Thermo Scientific Chemicals

L-Threonine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H62118.03
1 g, Each
Quantity
Catalog number: H62118.03
also known as H62118-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39994-75-7
IUPAC Name
methyl (2S,3R)-2-amino-3-hydroxybutanoate hydrochloride
Molecular Formula
C5H12ClNO3
InChI Key
OZSJLLVVZFTDEY-HJXLNUONSA-N
SMILES
Cl.COC(=O)[C@@H](N)[C@@H](C)O
Assay (unspecified)
>97.5%

Description

L-Threonine methyl ester hydrochloride is used as a feed and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Threonine methyl ester hydrochloride is used as a feed and food additive.

Solubility
Soluble in water.

Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text