Thermo Scientific Chemicals

D-Valine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H62131.14
25 g, Each
Thermo Scientific Chemicals

D-Valine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H62131.14
25 g, Each
Quantity
Catalog number: H62131.14
also known as H62131-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104944-18-5
IUPAC Name
tert-butyl (2R)-2-amino-3-methylbutanoate hydrochloride
Molecular Formula
C9H20ClNO2
InChI Key
AUIVQIHTTVPKFS-OGFXRTJISA-N
SMILES
Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C
Assay (unspecified)
>94.0%

Description

D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.

Notes
Store at -20°C. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text