Thermo Scientific Chemicals

Nalpha-Boc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%, Thermo Scientific Chemicals

Catalog number: H62148.06
5 g, Each
Thermo Scientific Chemicals

Nalpha-Boc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%, Thermo Scientific Chemicals

Catalog number: H62148.06
5 g, Each
Quantity
Catalog number: H62148.06
also known as H62148-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
200124-22-7
IUPAC Name
5-{[amino(2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonamido)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Molecular Formula
C24H38N4O7S
InChI Key
CVFXPOKENLGCID-UHFFFAOYNA-N
SMILES
CC1=C2CC(C)(C)OC2=C(C)C(C)=C1S(=O)(=O)NC(N)=NCCCC(NC(=O)OC(C)(C)C)C(O)=O
Assay (HPLC)
>95.0%

Description

Nalpha-Boc-Nμ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nµ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

Solubility
Soluble in water or 1% acetic acid

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text