Thermo Scientific Chemicals

trans-3-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H62207.03
1 g, Each
Thermo Scientific Chemicals

trans-3-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H62207.03
1 g, Each
Quantity
Catalog number: H62207.03
also known as H62207-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
218772-92-0
Appearance (Color)
Off-white to white
Form
Solid
Assay from Supplier's CofA
≥96.0%
Mass Spec
Conforms

Description

2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text