N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals
N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62210.03
also known as H62210-03
Price (USD)/ Each
78.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
78.80
Add to cart
N-Boc-D-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals
Catalog numberH62210.03
Price (USD)/ Each
78.80
-
Add to cart
Chemical Identifiers
CAS92828-64-3
IUPAC Name4-(benzyloxy)-3-{[(tert-butoxy)carbonyl]amino}-4-oxobutanoic acid
Molecular FormulaC16H21NO6
InChI KeyLDRWTKQWSXGSTM-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only