N-Boc-3,5-difluoro-L-phenylalanine, 95%, Thermo Scientific Chemicals
N-Boc-3,5-difluoro-L-phenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-3,5-difluoro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62216.06
also known as H62216-06
Price (USD)/ Each
590.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
590.00
Add to cart
N-Boc-3,5-difluoro-L-phenylalanine, 95%, Thermo Scientific Chemicals
Catalog numberH62216.06
Price (USD)/ Each
590.00
-
Add to cart
Chemical Identifiers
CAS205445-52-9
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,5-difluorophenyl)propanoic acid
Molecular FormulaC14H17F2NO4
InChI KeyCZBNUDVCRKSYDG-NSHDSACASA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC(F)=CC(F)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only