This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility Soluble in water, glycerol. Insoluble in ethanol, chloroform
Notes Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.