Nalpha-Fmoc-S-trityl-D-cysteine, 98%, Thermo Scientific Chemicals
Nalpha-Fmoc-S-trityl-D-cysteine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha-Fmoc-S-trityl-D-cysteine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62301.06
also known as H62301-06
Price (USD)/ Each
78.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
78.70
Add to cart
Nalpha-Fmoc-S-trityl-D-cysteine, 98%, Thermo Scientific Chemicals
Catalog numberH62301.06
Price (USD)/ Each
78.70
-
Add to cart
Chemical Identifiers
CAS167015-11-4
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular FormulaC37H31NO4S
InChI KeyKLBPUVPNPAJWHZ-UHFFFAOYNA-N
SMILESOC(=O)C(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only