Thermo Scientific Chemicals

DL-Prolinol, 98%, Thermo Scientific Chemicals

Catalog number: H62322.14
25 g, Each
Thermo Scientific Chemicals

DL-Prolinol, 98%, Thermo Scientific Chemicals

Catalog number: H62322.14
25 g, Each
Quantity
Catalog number: H62322.14
also known as H62322-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
498-63-5
IUPAC Name
(pyrrolidin-2-yl)methanol
Molecular Formula
C5H11NO
InChI Key
HVVNJUAVDAZWCB-UHFFFAOYNA-N
SMILES
OCC1CCCN1
Assay (GC)
>97.5%

Description

DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text