cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H62329.03
also known as H62329-03
Price (USD)/ Each
1,484.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
1,484.00
Add to cart
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH62329.03
Price (USD)/ Each
1,484.00
-
Add to cart
Specifications
Chemical Name or Materialcis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
CAS685508-28-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only