Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H62338.06
5 g, Each
Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H62338.06
5 g, Each
Quantity
Catalog number: H62338.06
also known as H62338-06
Price (USD)
Quantity
-
Have Questions?

Description

trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text