Nalpha-Boc-N^w-nitro-L-arginine, 98%, Thermo Scientific Chemicals
Nalpha-Boc-N^w-nitro-L-arginine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha-Boc-N^w-nitro-L-arginine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H62339.14
also known as H62339-14
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.90 
Save 13.25 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.90 
Save 13.25 (15%)
Add to cart
Nalpha-Boc-N^w-nitro-L-arginine, 98%, Thermo Scientific Chemicals
Catalog numberH62339.14
Price (USD)/ Each
76.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.90 
Save 13.25 (15%)
-
Add to cart
Chemical Identifiers
CAS2188-18-3
IUPAC Name(2S)-5-{[amino(nitroamino)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Molecular FormulaC11H21N5O6
InChI KeyOZSSOVRIEPAIMP-ZETCQYMHSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCN=C(N)N[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only