N-Boc-DL-phenylglycine, 98%, Thermo Scientific Chemicals
N-Boc-DL-phenylglycine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-DL-phenylglycine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62354.06
also known as H62354-06
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.10 
Save 9.45 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.10 
Save 9.45 (15%)
Add to cart
N-Boc-DL-phenylglycine, 98%, Thermo Scientific Chemicals
Catalog numberH62354.06
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.10 
Save 9.45 (15%)
-
Add to cart
Chemical Identifiers
CAS3601-66-9
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-2-phenylacetic acid
Molecular FormulaC13H17NO4
InChI KeyHOBFSNNENNQQIU-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only