Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H62373.03
1 g, Each
Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H62373.03
1 g, Each
Quantity
Catalog number: H62373.03
also known as H62373-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
215790-29-7
IUPAC Name
tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
Molecular Formula
C13H23NO3
InChI Key
OPGBSEXLSWYFOR-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC1CCC(CC=O)CC1
Appearance (Color)
White
Form
Solid
Proton NMR
Conforms to structure
Carbon-13 NMR
Conforms to structure
Assay (GC)
>97%

Description

trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

Solubility
Sparingly Soluble (0.41 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text