cis-4-Hydroxy-L-proline methyl ester hydrochloride, 95%, Thermo Scientific Chemicals
cis-4-Hydroxy-L-proline methyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-4-Hydroxy-L-proline methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H62384.03
also known as H62384-03
Price (USD)/ Each
238.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
238.00
Add to cart
cis-4-Hydroxy-L-proline methyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH62384.03
Price (USD)/ Each
238.00
-
Add to cart
Chemical Identifiers
CAS40126-30-5
IUPAC Namemethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride
Molecular FormulaC6H12ClNO3
InChI KeyKLGSHNXEUZOKHH-UHFFFAOYNA-N
SMILESCl.COC(=O)C1CC(O)CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.0% min
cis-4-Hydroxy-L-proline methyl ester hydrochloride is an Intermediate in pharmaceutical and organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Hydroxy-L-proline methyl ester hydrochloride is an Intermediate in pharmaceutical and organic syntheses.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
Jessica A. Smith; G. Paul Savage; and Oliver E. Hutt . Studies on the Synthesis of cis-4-Hydroxy-l-proline. Australian Journal of Chemistry. 2011, 64 (11), 1509-1514.