Thermo Scientific Chemicals

N-Fmoc-D-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H62395.14
25 g, Each
Thermo Scientific Chemicals

N-Fmoc-D-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H62395.14
25 g, Each
Quantity
Catalog number: H62395.14
also known as H62395-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79990-15-1
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular Formula
C18H17NO4
InChI Key
QWXZOFZKSQXPDC-UHFFFAOYNA-N
SMILES
CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>94.0%

Description

N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. It is also essential for the biosynthesis of peptidoglycan cross linking sub-units that are used for bacterial cell walls.

Solubility
Soluble in dimethylformamide (1 mole in 2 ml). Insoluble in water.

Notes
Refrigerate at 4°C. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text