Chemical Identifiers
CAS132388-68-2
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)carbamoyl]propanoic acid
Molecular FormulaC28H30N2O5
InChI KeyPYGOCFDOBSXROC-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
View more