Thermo Scientific Chemicals

trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals

Catalog number: H62469.MD
250 mg, Each
Thermo Scientific Chemicals

trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane, 97%, Thermo Scientific Chemicals

Catalog number: H62469.MD
250 mg, Each
Quantity
Catalog number: H62469.MD
also known as H62469-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1222709-30-9
IUPAC Name
tert-butyl N-[4-(bromomethyl)cyclohexyl]carbamate
Molecular Formula
C12H22BrNO2
InChI Key
JPJCPRFAUVDSEA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC1CCC(CBr)CC1
Appearance (Color)
White
Form
Solid
Carbon-13 NMR
Conforms to structure
Assay (GC)
>97%
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text