Thermo Scientific Chemicals

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H62472.06
5 g, Each
Thermo Scientific Chemicals

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H62472.06
5 g, Each
Quantity
Catalog number: H62472.06
also known as H62472-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14316-06-4
IUPAC Name
hydrogen methyl 2-aminopropanoate chloride
Molecular Formula
C4H10ClNO2
InChI Key
IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(C)N
Assay (Titration)
>97.5%

Description

D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.

Solubility
Soluble in Water 100 mg/ml.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text