Chemical Name or MaterialMethyl 4-bromo-2-fluorophenylacetate
CAS193290-19-6
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC9H8BrFO2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.