N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%, Thermo Scientific Chemicals
N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62495.06
also known as H62495-06
Price (USD)/ Each
672.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
672.00
Add to cart
N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH62495.06
Price (USD)/ Each
672.00
-
Add to cart
Chemical Identifiers
CAS89813-47-8
IUPAC Name2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular FormulaC17H23NO5
InChI KeyBEIPCYKSYYZEJH-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CC(O)CC1C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only