N-Boc-L-glutamic acid 5-methyl ester, 95%, Thermo Scientific Chemicals
N-Boc-L-glutamic acid 5-methyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-glutamic acid 5-methyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H62496.06
also known as H62496-06
Price (USD)/ Each
344.65
Online exclusive
383.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
344.65
Online exclusive
383.00 
Save 38.35 (10%)
Add to cart
N-Boc-L-glutamic acid 5-methyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH62496.06
Price (USD)/ Each
344.65
Online exclusive
383.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS45214-91-3
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-5-methoxy-5-oxopentanoic acid
Molecular FormulaC11H19NO6
InChI KeyOHYMUFVCRVPMEY-UHFFFAOYNA-N
SMILESCOC(=O)CCC(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only