Chemical Identifiers
CAS135610-90-1
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Molecular FormulaC40H33N3O4
InChI KeyXXMYDXUIZKNHDT-UHFFFAOYNA-N
SMILESOC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more