Thermo Scientific Chemicals

L-Tryptophanol, 97%

Catalog number: H62506.03
1 g, Each
Thermo Scientific Chemicals

L-Tryptophanol, 97%

Catalog number: H62506.03
1 g, Each
Quantity
Catalog number: H62506.03
also known as H62506-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2899-29-8
IUPAC Name
2-amino-3-(1H-indol-3-yl)propan-1-ol
Molecular Formula
C11H14N2O
InChI Key
UDQCRUSSQAXPJY-UHFFFAOYNA-N
SMILES
NC(CO)CC1=CNC2=CC=CC=C12
Assay (HPLC)
>96.0%

Description

L-Tryptophanol is an amino acid used in the construction of peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tryptophanol is an amino acid used in the construction of peptides and proteins.

Solubility
Soluble in water 11.4 g/L (25°C).

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text